Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Choline hydrogen tartrate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: CBT2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoateCholine bitartrate2-(Hydroxyethyl)trimethylammonium bitartrateCholine acid tartrate

J64657-A1 J64657-30 J64657-A4 J64657-30 J64657-A4 J64657-A1
AAJ64657-A1EA 133.9 USD InStock
CAAAJ64657-A4 AAJ64657-A1 AAJ64657-30 AAJ64657-A4 CAAAJ64657-A1 CAAAJ64657-30
Choline hydrogen tartrate 98+%
Choline hydrogen tartrate
Powder


Formula: C₉H₁₉NO₇
MW: 253.25 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00036332
CAS Number: 87-67-2
EINECS: 201-763-4
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty