Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Amino-1-(beta-D-ribofuranosyl)-1H-imidazole-4-carboxamideAcadesineAICAR

J64720-MF J64720-MF
AAJ64720-MFEA 169.4 USD InStock
CAAAJ64720-MF AAJ64720-MF
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 98%
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside
Activator of AMP-activated protein kinase (AMPK)

  • Powder

Powder


Formula: C₉H₁₄N₄O₅
MW: 258.23 g/mol
Melting Pt: 214…215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00869751
CAS Number: 2627-69-2
EINECS: 220-097-5
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty