Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tripeptidyl Peptidase Inhibitor

Supplier: Thermo Scientific
J64760-LB0 J64760-LB0 J64760-MA J64760-MA
AAJ64760-MAEA 204.94 USD InStock
AAJ64760-MA CAAAJ64760-MA AAJ64760-LB0 CAAAJ64760-LB0
Tripeptidyl Peptidase Inhibitor
Proteins and Peptides
A serine protease inhibitor that acts as a potent, selective, dose-dependent, irreversible inhibitor of tripeptidylpeptidase II

CAS No.: 184901-82-4
Molecular Formula: C16H22ClN3O3
Formula Weight: 453.90
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
MDL No.: MFCD01310850

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty