Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Supplier: Thermo Scientific Chemicals
Synonyms: Bz-Arg-pNA·HCl(S)-N-(5-Guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

J64915-ME J64915-MD J64915-MD J64915-MC J64915-MC J64915-ME
AAJ64915-MEEA 294.15 USD InStock
AAJ64915-ME CAAAJ64915-ME CAAAJ64915-MD CAAAJ64915-MC AAJ64915-MC AAJ64915-MD
N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride
Chromogenic trypsin substrate. Substrate for proteolytic enzymes

Powder

Applications: Chromogenic trypsin substrate. Substrate for proteolytic enzymes


Formula: C₁₉H₂₃ClN₆O₄
MW: 434.881 g/mol
Melting Pt: 223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063682
CAS Number: 21653-40-7
EINECS: 244-505-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty