Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Acetyl-L-serine hydrochloride

Supplier: Thermo Scientific Chemicals
Synonyms: H-Ser(Ac)-OH.HCl

J64933-06 J64933-03 J64933-03 J64933-06
AAJ64933-03EA 86.32 USD InStock
CAAAJ64933-03 AAJ64933-03 CAAAJ64933-06 AAJ64933-06
O-Acetyl-L-serine hydrochloride
O-Acetyl-L-serine hydrochloride
Substrate for cysteine synthase and beta-substituted alanine synthase in higher plants

Powder


Formula: C₅H₉NO₄
MW: 183.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00060169
CAS Number: 66638-22-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty