Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Deoxythymidine-5'-diphosphate trisodium salt 97%

Supplier: Thermo Scientific Chemicals
Warning

J64938-MC J64938-MB J64938-MC J64938-MB
AAJ64938-MCEA 349.01 USD InStock
AAJ64938-MC AAJ64938-MB CAAAJ64938-MC CAAAJ64938-MB
2'-Deoxythymidine-5'-diphosphate trisodium salt 97%
2'-Deoxythymidine-5'-diphosphate trisodium salt
Powder


Formula: C₃₀H₄₅N₆Na₃O₃₃P₆
MW: 1272.511 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00084748
CAS Number: 95648-78-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty