Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Native Avidin (from Egg)

Supplier: Thermo Scientific
J64972-MA J64972-MA
AAJ64972-MAEA 154.16 USD InStock
AAJ64972-MA CAAAJ64972-MA
Native Avidin (from Egg)
Proteins and Peptides
Lyophilized powder

CAS No.: 1405-69-2
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD00130572

Applications: Has extremely strong affinity for biotin

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty