Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cromolyn sodium 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Disodium Cromoglycate5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis(4-oxo-4H-1-benzopyran-2-carboxylic Acid) Disodium Salt

J65122-06 J65122-03 J65122-06 J65122-14 J65122-03 J65122-14
AAJ65122-06EA 215.01 USD InStock
AAJ65122-06 AAJ65122-03 AAJ65122-14 CAAAJ65122-06 CAAAJ65122-03 CAAAJ65122-14
Cromolyn sodium 98%
Cromolyn sodium
Mast cell stabilizer that prevents release of histamine from mast cells

Powder


Formula: C₂₃H₁₆O₁₁
MW: 512.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057744
CAS Number: 15826-37-6
EINECS: 239-926-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty