Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ɑ-Asarone

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: trans-1,2,4-Trimethoxy-5-(1-propenyl)benzene

J65125-06 J65125-06 J65125-03 J65125-03
AAJ65125-06EA 203.92 USD InStock
CAAAJ65125-03 AAJ65125-06 AAJ65125-03 CAAAJ65125-06
ɑ-Asarone
alpha-Asarone
Powder


Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Boiling Pt: 296 °C
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00064457
CAS Number: 2883-98-9
EINECS: 220-743-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty