Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Compound E, solid

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-2-(2-(3,5-Difluorophenyl)acetamido)-N-((S)-1-methyl-2-oxo-5-phenyl-2,3-dihydro-1H-benzo[e][1,4]diazepin-3-yl)propanamide

J65131-EXD J65131-EXD
AAJ65131-EXDEA 321.59 USD InStock
CAAAJ65131-EXD AAJ65131-EXD
Compound E, solid
Compound E
A cell-permeable, potent, selective, peptidomimetic, non-transition-state analog inhibitor of -secretase and Notch processing


Formula: C₂₇H₂₄F₂N₄O₃
MW: 490.511 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04974531
CAS Number: 209986-17-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty