Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-hydroxybutyric acid

Supplier: Thermo Scientific Chemicals

J65145-22 J65145-06 J65145-22 J65145-14 J65145-06 J65145-14
AAJ65145-06EA 65.74 USD InStock
AAJ65145-06 CAAAJ65145-22 CAAAJ65145-14 AAJ65145-14 CAAAJ65145-06 AAJ65145-22
4-Amino-3-hydroxybutyric acid
4-Amino-3-hydroxybutyric acid
Reduces uptake of iodine by the thyroid glands in animals. Antispasmodic

Powder


Formula: C₄H₉NO₃
Melting Pt: 223 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008141
CAS Number: 924-49-2
EINECS: 213-106-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty