Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2'-Deoxyguanosine-5'-monophosphoric acid disodium salt

J65186-14 J65186-03 J65186-03 J65186-06 J65186-06 J65186-14
AAJ65186-06EA 299.24 USD InStock
CAAAJ65186-06 CAAAJ65186-03 CAAAJ65186-14 AAJ65186-06 AAJ65186-14 AAJ65186-03
2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate 98%
2'-Deoxyguanosine-5'-monophosphate disodium salt hydrate
Crystalline powder


Formula: C₁₀H₁₂N₅Na₂O₇P.xH₂O
MW: 391.18 g/mol (anhy)
Melting Pt: >245 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00080331
CAS Number: 33430-61-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty