Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human;Bovine Orexin A

Supplier: Thermo Scientific
J65378-EXD J65378-EXD
AAJ65378-EXDEA 595.92 USD InStock
CAAAJ65378-EXD AAJ65378-EXD
Human;Bovine Orexin A
Proteins and Peptides
Molecular Formula: C152H243N47O44S4
Formula Weight: 3561.09
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Solubility: Soluble in water at 1mg/ml
MDL No.: MFCD07369024

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty