Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Deoxy-D-galactose 99%

Supplier: Thermo Scientific Chemicals

J65482-09 J65482-04 J65482-04 J65482-06 J65482-09 J65482-06
AAJ65482-09EA 583.77 USD InStock
AAJ65482-09 CAAAJ65482-09 CAAAJ65482-06 CAAAJ65482-04 AAJ65482-04 AAJ65482-06
2-Deoxy-D-galactose 99%
2-Deoxy-D-galactose
For studying galactose uptake into Escherichia coli

Powder


Formula: C₆H₁₂O₅
MW: 164.16 g/mol
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014649
CAS Number: 1949-89-9
EINECS: 217-765-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty