Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aza-2'-deoxycytidine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosineDecitabine

J65513-MD J65513-MD J65513-MC J65513-MC
AAJ65513-MCEA 294.18 USD InStock
CAAAJ65513-MD AAJ65513-MC AAJ65513-MD CAAAJ65513-MC
5-Aza-2'-deoxycytidine 98%
5-Aza-2'-deoxycytidine
Powder


Formula: C₈H₁₂N₄O₄
MW: 228.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043011
CAS Number: 2353-33-5
EINECS: 219-089-4
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty