Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Bromoadenine

Supplier: Thermo Scientific Chemicals
Synonyms: 8-Bromo-1H-adenine

J65582-MC J65582-ME J65582-ME J65582-03 J65582-MC J65582-03
AAJ65582-MEEA 226.24 USD InStock
CAAAJ65582-03 CAAAJ65582-MC AAJ65582-ME AAJ65582-MC AAJ65582-03 CAAAJ65582-ME
8-Bromoadenine
8-Bromoadenine
Powder


Formula: C₅H₄BrN₅
MW: 214.02 g/mol
Melting Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00082518
CAS Number: 6974-78-3
EINECS: 230-225-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty