Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pig Dynorphin A amide

Supplier: Thermo Scientific
J65622-MCR J65622-MCR
AAJ65622-MCREA 109.54 USD InStock
AAJ65622-MCR CAAAJ65622-MCR
Pig Dynorphin A amide
Proteins and Peptides
CAS No.: 80448-90-4
Molecular Formula: C99H156N32O22
Formula Weight: 2146.49
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00076351

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty