Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Laminin A Chain (2091-2108)

Supplier: Thermo Scientific
J65640-MCR J65640-MCR J65640-EXD J65640-EXD
AAJ65640-MCREA 226.23 USD InStock
CAAAJ65640-EXD AAJ65640-MCR AAJ65640-EXD CAAAJ65640-MCR
Laminin A Chain (2091-2108)
Proteins and Peptides
Molecular Formula: C82H148N31O26S
Formula Weight: 2016.30
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty