Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-histidine monohydrate

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-His-OH

J65657-06 J65657-14 J65657-03 J65657-14 J65657-03 J65657-06
AAJ65657-14EA 444.37 USD InStock
AAJ65657-14 AAJ65657-03 AAJ65657-06 CAAAJ65657-03 CAAAJ65657-14 CAAAJ65657-06
N-Acetyl-L-histidine monohydrate
N-Acetyl-L-histidine monohydrate
Powder


Formula: C₈H₁₁N₃O₃
MW: 215.2 g/mol
Melting Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00149320
CAS Number: 39145-52-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty