Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cytosine-β-D-arabinofuranoside hydrochloride

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Arabinosylcytosine hydrochloride1-(beta-D-Arabinofuranosyl)cytosine HClCytarabine HCl

J65671-09 J65671-09 J65671-06 J65671-03 J65671-06 J65671-03
AAJ65671-06EA 112.59 USD InStock
AAJ65671-06 CAAAJ65671-09 AAJ65671-09 AAJ65671-03 CAAAJ65671-03 CAAAJ65671-06
Cytosine-β-D-arabinofuranoside hydrochloride
Cytosine-beta-D-arabinofuranoside hydrochloride
Interferes with the synthesis of DNA

Powder


Formula: C₉H₁₄ClN₃O₅
MW: 279.677 g/mol
Melting Pt: 197…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00012839
CAS Number: 69-74-9
EINECS: 200-713-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty