Synonyms:
ADIBO-PEG4-maleimide
J65673-MB
J65673-MC
J65673-MA
J65673-MA
J65673-MB
J65673-MC
AAJ65673-MBEA
318.52
USD
InStock
AAJ65673-MB
AAJ65673-MA
AAJ65673-MC
CAAAJ65673-MB
CAAAJ65673-MA
CAAAJ65673-MC
Azadibenzocyclooctyne-PEG4-maleimide
Azadibenzocyclooctyne-PEG4-maleimide
A sulfhydryl reactive reagent with PEG4 spacer for incorporating ADIBO into molecules
Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. The PEG spacer adds hydrophilicity to the product to decrease aggregation.
Caution: Notes: Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.