Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Deoxycytidine-5'-monophosphate disodium salt

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium ((2R,3S,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxytetrahydrofuran-2-yl)methyl phosphate

J65724-03 J65724-03
AAJ65724-03EA 116.65 USD InStock
CAAAJ65724-03 AAJ65724-03
2'-Deoxycytidine-5'-monophosphate disodium salt
2'-Deoxycytidine-5'-monophosphate disodium salt
Powder


Formula: C₉H₁₂N₃Na₂O₇P
MW: 351.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00069774
CAS Number: 13085-50-2
EINECS: 235-995-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty