Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human [Des-octanoyl]-Ghrelin

Supplier: Thermo Scientific
J65946-MCR J65946-EXD J65946-MCR J65946-EXD
AAJ65946-MCREA 342.9 USD InStock
AAJ65946-MCR AAJ65946-EXD CAAAJ65946-EXD CAAAJ65946-MCR
Human [Des-octanoyl]-Ghrelin
Proteins and Peptides
CAS No.: 313951-59-6
Molecular Formula: C141H235N47O41
Formula Weight: 3244.66
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD08460342

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty