Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl-L-aspartate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-Aminosuccinic Acid Dimethyl Ester HydrochlorideL-Asparaginic Acid Dimethyl Ester HydrochlorideDimethyl (S)-Aminosuccinate HydrochlorideDimethyl L-Asparaginate HydrochlorideL-Aspartic Acid Dimethyl Ester HydrochlorideH-Asp(OMe)-OMe.HCl

J65983-06 J65983-06 J65983-14 J65983-14
AAJ65983-06EA 79.84 USD InStock
AAJ65983-06 CAAAJ65983-06 CAAAJ65983-14 AAJ65983-14
Dimethyl-L-aspartate hydrochloride 98%
Dimethyl-L-aspartate hydrochloride
Powder


Formula: C₆H₁₁NO₄
MW: 197.62 g/mol
Melting Pt: 115…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00038878
CAS Number: 32213-95-9
EINECS: 250-957-4
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty