Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fibrinogen Related Peptide

Supplier: Thermo Scientific
J66069-MA J66069-LB0
AAJ66069-MAEA 567.13 USD InStock
AAJ66069-MA AAJ66069-LB0
Fibrinogen Related Peptide
Proteins and Peptides
Fibrinogen Related Peptide

CAS No.: 80755-86-8
Molecular Formula: C62H99N23O21
Formula Weight: 1502.59
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00133316

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty