Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cortistatin 14

Supplier: Thermo Scientific
J66255-MCR J66255-EXD
AAJ66255-EXDEA 116.66 USD InStock
AAJ66255-EXD AAJ66255-MCR
Cortistatin 14
Proteins and Peptides
Cortistatin 14

CAS: 186901-48-4
Form: Solid
MDL: MFCD01075120

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty