Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Deoxynojirimycin hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3R,4R,5S)-2-(Hydroxymethyl)piperidine-3,4,5-triol hydrochloride

J66458-MA J66458-MB
AAJ66458-MAEA 164.32 USD InStock
AAJ66458-MA 10814-658
1-Deoxynojirimycin hydrochloride 98%
1-Deoxynojirimycin hydrochloride
White powder

Soluble in water at 20 mg/mL


Formula: C₆H₁₃NO₄·HCl
MW: 199.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00133247
CAS Number: 73285-50-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty