Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt

Supplier: Thermo Scientific Chemicals

J66461-EXD J66461-EXZ
AAJ66461-EXZEA 109.56 USD InStock
AAJ66461-EXZ AAJ66461-EXD
D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt
D-myo-Inositol-1,3,4,5-tetrakis(phosphate) octasodium salt
Lyophilized powder


Formula: C₆H₈Na₈O₁₈P₄
MW: 675.925 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00153818
CAS Number: 210488-61-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty