Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Urocortin III

Supplier: Thermo Scientific
J66478-MCR
AAJ66478-MCREA 330.72 USD InStock
AAJ66478-MCR
Mouse Urocortin III
Proteins and Peptides
Urocortin III, mouse

CAS: 357952-10-4
Form: Solid
MDL: MFCD03792654

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty