Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat Adrenocorticotropic Hormone (1-39)

Supplier: Thermo Scientific
J66519-MCR J66519-EXD
AAJ66519-EXDEA 310.48 USD InStock
AAJ66519-EXD AAJ66519-MCR
Rat Adrenocorticotropic Hormone (1-39)
Proteins and Peptides
Adrenocorticotropic Hormone (1-39), rat

CAS: 77465-10-2
Form: Solid
MDL: MFCD00167431

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty