Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide, Bioreagent

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DMSO

J66650-K2 J66650-AD J66650-AP J66650-AE J66650-AK
AAJ66650-APEA 186.36 USD InStock
AAJ66650-AP AAJ66650-AK AAJ66650-K2 AAJ66650-AE AAJ66650-AD
Dimethyl sulfoxide, Bioreagent
Dimethyl sulphoxide
Suitable for biotechnology applications such as peptide and oligonucleotide synthesis

Liquid

This product is not sterile.


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C
Melting Pt: 18.4 °C
Density: 1.101
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty