Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cholecystokinin (26-33)

Supplier: Thermo Scientific
J66669-EXD J66669-MCR
AAJ66669-EXDEA 112.78 USD InStock
AAJ66669-EXD AAJ66669-MCR
Cholecystokinin (26-33)
Proteins and Peptides
Cholecystokinin, CCK Octapeptide (26-33)

CAS: 25126-32-3
Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty