Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proinsulin C-Peptide (55-89), human

Supplier: Thermo Scientific
J66768-MCR
AAJ66768-MCREA 340.89 USD InStock
AAJ66768-MCR
Proinsulin C-Peptide (55-89), human
Proteins and Peptides
Proinsulin C-Peptide (55-89), human

CAS: 11097-48-6
Form: Solid
MDL: MFCD00076294

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty