Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neurokinin A

Supplier: Thermo Scientific
J66817-EXD J66817-MCR
AAJ66817-EXDEA 98.58 USD InStock
AAJ66817-EXD AAJ66817-MCR
Neurokinin A
Proteins and Peptides
Neurokinin A

CAS: 86933-74-6
Form: Powder
MDL: MFCD00167560

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty