Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Selenomethionine

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S)-2-Amino-4-(methylseleno)butyric Acid

J66902-03 J66902-MC J66902-ME
AAJ66902-03EA 665.45 USD InStock
AAJ66902-03 AAJ66902-ME AAJ66902-MC
L-Selenomethionine
L-Selenomethionine
Seleno-L-methionine


Formula: C₅H₁₁NO₂Se
MW: 196.11 g/mol
Melting Pt: 265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037210
CAS Number: 3211-76-5
UN: 3283
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty