Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bradykinin (1-6)

Supplier: Thermo Scientific
J66937-LB0 J66937-MB
AAJ66937-MBEA 224.51 USD InStock
AAJ66937-MB AAJ66937-LB0
Bradykinin (1-6)
Proteins and Peptides
Bradykinin (1-6)

CAS No.: 23815-88-5
Molecular Formula: C30H45N9O8
Formula Weight: 659.73
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00076261

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty