Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

17-AAG 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 17-(Allylamino)-17-demethoxygeldanamycin(4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

J66960-EX3 J66960-MB J66960-MC
AAJ66960-EX3EA 426.04 USD InStock
AAJ66960-EX3 AAJ66960-MC AAJ66960-MB
17-AAG 99%
17-AAG
17-(Allylamino)-17-demethoxygeldanamycin (17-AAG) inhibits heat shock protein 90 (Hsp90), the expression of heat shock factor-1, and the activity of oncogenic proteins such as N-ras, Ki-ras, c-Akt, and p185erB2

  • Possesses anti-tumor activity

17-AAG also induces apoptosis alond with its anti-tumor effects.


Formula: C₃₁H₄₃N₃O₈
MW: 585.7 g/mol
Melting Pt: 207…209 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD04973892
CAS Number: 75747-14-7
Order Now
Specification Test Results
Assay 99%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty