Synonyms:
17-(Allylamino)-17-demethoxygeldanamycin
, (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
J66960-EX3
J66960-MB
J66960-MC
AAJ66960-EX3EA
426.04
USD
InStock
AAJ66960-EX3
AAJ66960-MC
AAJ66960-MB
17-AAG 99%
17-AAG
17-(Allylamino)-17-demethoxygeldanamycin (17-AAG) inhibits heat shock protein 90 (Hsp90), the expression of heat shock factor-1, and the activity of oncogenic proteins such as N-ras, Ki-ras, c-Akt, and p185erB2
- Possesses anti-tumor activity
17-AAG also induces apoptosis alond with its anti-tumor effects.