Would you like to log in again?
DNA Molecular Weight Marker, Wide Range Ladder, Thermo Scientific
Supplier: Thermo Scientific
J67051-AMI
AAJ67051-AMIEA 302.33 USD InStock
AAJ67051-AMI
DNA Molecular Weight Marker, Wide Range Ladder, Thermo Scientific
Electrophoresis Reagents Molecular Weight Markers
This DNA Molecular Weight Marker is used for rapid and precise sizing of DNA strands.
The bands appear in 100 bp increments from 100 to 1,000 bp. The bands are in 1,000 bp increments from 1 to 12 kb. Bands at 1 kb and 5 kb stain brighter to aid identification.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty