Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Prostaglandin F2α tris salt 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol (Z)-7-((1R,2R,3R,5S)-3,5-Dihydroxy-2-((S,E)-3-hydroxyoct-1-en-1-yl)cyclopentyl)hept-5-enoate

J67245-LB0 J67245-MA J67245-MCR
AAJ67245-MAEA 578.45 USD InStock
AAJ67245-MA AAJ67245-MCR AAJ67245-LB0
Prostaglandin F2α tris salt 99%
Prostaglandin F2alpha tris salt
Solid


Formula: C₂₄H₄₅NO₈
MW: 475.623 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077863
CAS Number: 38562-01-5
EINECS: 254-002-3
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty