Would you like to log in again?
Krebs-Ringer Solution, Bicarbonate-Buffered, Thermo Scientific
Supplier: Thermo Scientific
J67591-AP J67591-K2
AAJ67591-K2EA 174.97 USD InStock
AAJ67591-K2 AAJ67591-AP
Krebs-Ringer Solution, Bicarbonate-Buffered, Thermo Scientific
Buffers Cell Culture Buffers
Contains: 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5.5 mM D-glucose, pH 7.3. Prepared in 18.2 megohms water. Filtered through 0.22 micron filter, rendering it virtually sterile, but it is not tested for sterility and cannot be certified as such.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty