Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ɑ-Cyano-4-hydroxycinnamic acid, ultrapure matrix substance for MALDI

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Cyano-3-(4-hydroxyphenyl)acrylic Acidalpha-CHCA

J67635-EXK J67635-EXK
AAJ67635-EXKEA 182.46 USD InStock
AAJ67635-EXK CAAAJ67635-EXK
ɑ-Cyano-4-hydroxycinnamic acid, ultrapure matrix substance for MALDI
alpha-Cyano-4-hydroxycinnamic acid
Applications: For use as a matrix in MALDI mass spectrometry analysis


Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 250…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00004204
CAS Number: 28166-41-8
EINECS: 248-879-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty