Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hanks' Balanced Salt Solution (1X), with Calcium, with Magnesium, with Phenol Red, Thermo Scientific

Supplier: Thermo Scientific
J67799-K2 J67799-AP
AAJ67799-K2EA 140.19 USD InStock
AAJ67799-K2 AAJ67799-AP
Hanks' Balanced Salt Solution (1X), with Calcium, with Magnesium, with Phenol Red, Thermo Scientific
Buffers Cell Culture Buffers
Physical Form: Liquid
Appearance: Red
MDL No.: MFCD00217418

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty