Would you like to log in again?
Krebs-Ringer Solution, without Calcium, Thermo Scientific
Supplier: Thermo Scientific
J67839-AP J67839-K2
AAJ67839-APEA 133.79 USD InStock
AAJ67839-AP AAJ67839-K2
Krebs-Ringer Solution, without Calcium, Thermo Scientific
Buffers Cell Culture Buffers
Contains: 120 mM sodium chloride, 5 mM potassium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5.5 mM D-glucose, pH 7.3. Prepared in 18.2 megohms water and filtered through 0.22 micron filter.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty