Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Butanediol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,4-Butylene glycol1,4-Dihydroxy butaneTetramethylene glycolBDO

L03491-30 L03491-0E L03491-0B
AAL03491-0EEA 76.56 USD InStock
AAL03491-0E AAL03491-0B AAL03491-30
1,4-Butanediol 99%
1,4-Butanediol


Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 229…230 °C
Melting Pt: 19…20 °C
Density: 1.017
Flash Pt: 135 °C(275 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002968
CAS Number: 110-63-4
EINECS: 203-786-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty