Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydriodic acid 57% (w/w) in aqueous solution stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydrogen iodideHydroiodic acid

Stabilization: stab. with hypophosphorous acid 1.5%

L10410-22 L10410-36
AAL10410-22EA 80 USD InStock
AAL10410-22 AAL10410-36
Hydriodic acid 57% (w/w) in aqueous solution stabilized
Hydriodic acid


Formula: HI
MW: 127.91 g/mol
Boiling Pt: 126…127 °C
Density: 1.701
Storage Temperature: Ambient
MDL Number: MFCD00011347
CAS Number: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty