Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-Prolinol 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: D-ProlinolD(-)-Prolinol(2R)-pyrrolidin-2-ylmethanol

L11109-14 L11109-03 L11109-06 L11109-03
AAL11109-06EA 278.99 USD InStock
CAAAAL11109-03 AAL11109-06 AAAL11109-03 AAL11109-14
(R)-(-)-Prolinol 98+%
(R)-(-)-Prolinol


Formula: C₅H₁₁NO
MW: 101.1 g/mol
Boiling Pt: 74…76 °C/2 mm
Density: 1.025
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064321
CAS Number: 68832-13-3
EINECS: 629-147-4
Order Now
Specification Test Results
Optical rotation

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty