Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Trimethylsilylethynyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-((Trimethylsilyl)ethynyl)pyridine2-[(Trimethylsilyl)ethynyl]pyridine(2-Pyridylethynyl)trimethylsilane

L11251-03 L11251-06
AAL11251-06EA 186.99 USD InStock
AAL11251-06 AAL11251-03
2-(Trimethylsilylethynyl)pyridine 97%
2-(Trimethylsilylethynyl)pyridine


Formula: C₁₀H₁₃NSi
MW: 175.31 g/mol
Boiling Pt: 85…86 °C/1 mm
Melting Pt: 40…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00066349
CAS Number: 86521-05-3
EINECS: 000-000-0
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty