Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1H-Indene

L12665-14 L12665-06 L12665-22 L12665-14 L12665-06 L12665-22
AAAL12665-06EA 72.25 USD InStock
CAAAL12665-22 AAAL12665-06 CAAAAL12665-06 AAL12665-22 CAAAAL12665-14 AAAL12665-14
Indene 97%
Indene


Formula: C₉H₈
MW: 116.16 g/mol
Boiling Pt: 181…182 °C
Melting Pt: ca …2 °C
Density: 0.996
Flash Pt: 58 °C(136 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003777
CAS Number: 95-13-6
EINECS: 202-393-6
UN: 3295
ADR: "3",III
Merck Index: 13,04964
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty