Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-ethylenediamine 98% may cont up to 5% tert-butanol

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: t-boc-N-EDAtert-Butyl N-(2-Aminoethyl)carbamateN-(tert-Butoxycarbonyl)-1,2-ethylenediaminetert-Butyl (2-aminoethyl)carbamateN-(tert-Butoxycarbonyl)-1,2-diaminoethaneN-BOC-1,2-diaminoethane

L19947-06 L19947-06 L19947-14
AAAL19947-06EA 124.99 USD InStock
AAAL19947-06 CAAAAL19947-06 AAL19947-14
N-Boc-ethylenediamine 98% may cont up to 5% tert-butanol
N-Boc-ethylenediamine


Formula: C₇H₁₆N₂O₂
MW: 160.22 g/mol
Boiling Pt: 72 °C/0.1 mm
Density: 1.016
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
EINECS: 000-000-0
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty